Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432686_m1
          See other CSF1 GE Assays ›
          Gene Symbol:
          Csf1
          Gene Name:
          colony stimulating factor 1 (macrophage)
          Gene Aliases:
          C87615, Csfm, MCSF, op
          Chromosome Location:
          Chr.3: 107741048 - 107760469 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432686_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113529.1 NP_001107001.1 6 - 7 1081 70
          NM_001113530.1 NP_001107002.1 6 - 7 1966 70
          NM_007778.4 NP_031804.3 6 - 7 1966 70
          GenBank mRNA AK138489.1 - 6 - 7 1740 70
          AK154261.1 - 6 - 7 70
          AK154872.1 - 6 - 7 1740 70
          AK160995.1 - 6 - 7 1740 70
          AK166370.1 - 6 - 7 1739 70
          AK170154.1 - 6 - 7 1676 70
          BC025593.1 - 6 - 7 867 70
          BC066187.1 - 6 - 7 1720 30106218 70
          BC066200.1 - 6 - 7 1720 30100524 70
          BC066205.1 - 6 - 7 1720 30100601 70
          M21149.1 - 6 - 7 1629 70
          M21952.1 - 6 - 7 1719 70
          S78392.1 - 6 - 7 834 70
          X05010.1 - 6 - 7 1741 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002718.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csf1

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          positive regulation of cell-matrix adhesion
          osteoclast proliferation
          immune system process
          developmental process involved in reproduction
          inflammatory response
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell proliferation
          positive regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of macrophage chemotaxis
          macrophage differentiation
          regulation of ossification
          osteoclast differentiation
          positive regulation of cell migration
          positive regulation of cellular protein metabolic process
          positive regulation of mononuclear cell proliferation
          macrophage colony-stimulating factor signaling pathway
          positive regulation of multicellular organism growth
          odontogenesis
          positive regulation of odontogenesis of dentin-containing tooth
          innate immune response
          positive regulation of macrophage differentiation
          positive regulation of monocyte differentiation
          positive regulation of osteoclast differentiation
          positive regulation of protein kinase activity
          positive regulation of Ras protein signal transduction
          homeostasis of number of cells within a tissue
          branching involved in mammary gland duct morphogenesis
          mammary gland fat development
          mammary duct terminal end bud growth
          positive regulation of macrophage colony-stimulating factor signaling pathway
          positive regulation of microglial cell migration
          cytokine activity
          macrophage colony-stimulating factor receptor binding
          protein binding
          growth factor activity
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 NM_001113529.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 NM_001113529.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 NM_001113530.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 NM_001113530.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 NM_007778.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 NM_007778.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021181.mm.1 TC0300002718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021184.mm.1 NM_001113530.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021184.mm.1 NM_001113530.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021184.mm.1 NM_007778.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021184.mm.1 NM_007778.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021184.mm.1 TC0300002718.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 299973 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline