Search
Search

Gabrd| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008072.2 | NP_032098.2 | 2 - 3 | 288 | 74 | |
| GenBank mRNA | BC116832.1 | - | 2 - 3 | 223 | 40126151 | 74 |
| BC116836.1 | - | 2 - 3 | 223 | 40126155 | 74 | |
| BY276433.1 | - | 2 - 3 | 332 | 74 | ||
| S42882.1 | - | 2 - 3 | 281 | 74 | ||
Set Membership: |
|