Search
Search
| Gene Symbol: Grn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008175.4 | NP_032201.2 | 11 - 12 | 1635 | 68 | |
| GenBank mRNA | AK013678.1 | - | 11 - 12 | 1451 | 68 | |
| AK018744.1 | - | 11 - 12 | 1476 | 68 | ||
| AK147932.1 | - | 11 - 12 | 1452 | 68 | ||
| AK149570.1 | - | 11 - 12 | 1454 | 68 | ||
| AK150205.1 | - | 11 - 12 | 1451 | 68 | ||
| AK150338.1 | - | 11 - 12 | 1440 | 68 | ||
| AK150420.1 | - | 11 - 12 | 1439 | 68 | ||
| AK150717.1 | - | 11 - 12 | 1451 | 68 | ||
| AK150746.1 | - | 11 - 12 | 1453 | 68 | ||
| AK150881.1 | - | 11 - 12 | 1453 | 68 | ||
| AK151007.1 | - | 11 - 12 | 1451 | 68 | ||
| AK151162.1 | - | 11 - 12 | 1453 | 68 | ||
| AK151343.1 | - | 11 - 12 | 1451 | 68 | ||
| AK151713.1 | - | 11 - 12 | 1451 | 68 | ||
| AK151756.1 | - | 11 - 12 | 1451 | 68 | ||
| AK152048.1 | - | 11 - 12 | 1451 | 68 | ||
| AK152087.1 | - | 11 - 12 | 1451 | 68 | ||
| AK152118.1 | - | 11 - 12 | 1440 | 68 | ||
| AK152213.1 | - | 11 - 12 | 1440 | 68 | ||
| AK152705.1 | - | 11 - 12 | 1451 | 68 | ||
| AK152765.1 | - | 11 - 12 | 1451 | 68 | ||
| AK152944.1 | - | 11 - 12 | 1440 | 68 | ||
| AK153183.1 | - | 11 - 12 | 1441 | 68 | ||
| AK153289.1 | - | 11 - 12 | 1451 | 68 | ||
| AK153418.1 | - | 11 - 12 | 1451 | 68 | ||
| AK153470.1 | - | 11 - 12 | 1451 | 68 | ||
| AK154492.1 | - | 11 - 12 | 1451 | 68 | ||
| AK159397.1 | - | 11 - 12 | 1440 | 68 | ||
| AK159428.1 | - | 11 - 12 | 1440 | 68 | ||
| AK159557.1 | - | 11 - 12 | 1451 | 68 | ||
| AK159809.1 | - | 11 - 12 | 1451 | 68 | ||
| AK160022.1 | - | 11 - 12 | 1440 | 68 | ||
| AK160033.1 | - | 11 - 12 | 1451 | 68 | ||
| AK160074.1 | - | 11 - 12 | 1440 | 68 | ||
| BC037047.1 | - | 10 - 11 | 1432 | 68 | ||
| BC049943.1 | - | 11 - 12 | 1437 | 68 | ||
| BC129849.1 | - | 11 - 12 | 1435 | 40109630 | 68 | |
| BC129850.1 | - | 11 - 12 | 1435 | 40109631 | 68 | |
| M86736.1 | - | 11 - 12 | 1433 | 68 | ||
| X62321.1 | - | 10 - 11 | 1416 | 68 | ||
Set Membership: |
|