Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438315_s1
          See other PTGDR2 GE Assays ›
          Gene Symbol:
          Ptgdr2
          Gene Name:
          prostaglandin D2 receptor 2
          Gene Aliases:
          Crth2, Gpr44, Grp45
          Chromosome Location:
          Chr.19: 10927861 - 10942511 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438315_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptgdr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009962.3 NP_034092.1 3 305 58
          GenBank mRNA AB109092.1 - 3 305 58
          AK041947.1 - 3 303 58
          BC111906.1 - 3 285 40039170 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000213.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptgdr2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          calcium-mediated signaling
          positive regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of male germ cell proliferation
          prostaglandin J receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          prostaglandin D receptor activity
          prostaglandin F receptor activity
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953767 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953769 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953770 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2130463 AK036812.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2130463 NM_023731.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2130463 AB109092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2130463 NM_009962.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72060 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176797 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2130463 AK036812.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2130463 NM_023731.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2130463 AB109092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2130463 NM_009962.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P593486 AK036812.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P593486 NM_023731.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P593486 AB109092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P593486 AK041947.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P593486 NM_009962.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P406315 BC111906.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P406315 AB109092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P406315 NM_009962.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953760 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09105292 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176797 - -
          Whole Mouse Genome G4122A A_52_P593486 AK036812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593486 NM_023731.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593486 AB109092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593486 AK041947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593486 NM_009962.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406315 BC111906.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406315 AB109092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406315 NM_009962.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134740 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176797 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14313285 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14313277 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14313281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14313280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953769 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14313289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14252213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14252211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14454596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14313273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14193377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14422879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14467697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14467703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14252209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14313277 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461586 - -
          Affymetrix HT MG-430 PM Array Plate 1422275_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461583 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466104 - -
          Mouse Genome 430 2.0 Array 1422275_at - -
          Mouse Genome 430 2.0 Array 1422275_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14764_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14764_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002684.mm.1 NM_009962.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002684.mm.1 TC1900000213.mm.1 MTA TC
          Mouse Expression Set 430 1422275_PM_at - -
          Mouse Expression Set 430 1422275_at - -
          Murine Genome U74v2 Set 101282_at - -
          Murine Genome U74v2 Set 101282_PM_at - -
          Mouse Genome 430A 2.0 Array 1422275_at - -
          Mouse Genome 430A 2.0 Array 1422275_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422275_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422275_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 579271 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4050452 - -
          Illumina mouse-6 v1.0 expression beadchip 4050452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234360 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234360 - -
          mouse-6 v1.1 (Illumina) scl53364.3_607-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53364.3_607-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234360 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234360 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline