Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439085_m1
          See other GFRA1 GE Assays ›
          Gene Symbol:
          Gfra1
          Gene Name:
          glial cell line derived neurotrophic factor family receptor alpha 1
          Gene Aliases:
          AU042498
          Chromosome Location:
          Chr.19: 58235581 - 58455946 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439085_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gfra1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010279.3 NP_034409.1 1 - 2 197 80
          NR_104342.1 1 - 2 197 80
          GenBank mRNA AK076171.1 - 1 - 2 117 80
          BC044783.2 - 1 - 2 76 80
          BY233103.1 - 1 - 2 198 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001722.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_010279.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gfra1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane receptor protein tyrosine kinase signaling pathway
          nervous system development
          anatomical structure morphogenesis
          cell migration
          neuron differentiation
          neuron projection development
          receptor activity
          integrin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900015259.mm.1 NM_001285457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015259.mm.1 NM_010279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015259.mm.1 NR_104342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015259.mm.1 TC1900001722.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900015265.mm.1 NM_010279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015265.mm.1 NR_104342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015265.mm.1 TC1900001722.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900015264.mm.1 NM_010279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015264.mm.1 NR_104342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015264.mm.1 TC1900001722.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900015266.mm.1 NM_010279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015266.mm.1 NR_104342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015266.mm.1 TC1900001722.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900015258.mm.1 NM_001285457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015258.mm.1 NM_010279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015258.mm.1 NR_104342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015258.mm.1 TC1900001722.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900015267.mm.1 NM_010279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015267.mm.1 NR_104342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015267.mm.1 TC1900001722.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906691 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline