Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439226_m1
          See other H2-DMA GE Assays ›
          Gene Symbol:
          H2-DMa
          Gene Name:
          histocompatibility 2, class II, locus DMa
          Gene Aliases:
          H-2Ma, H2-Ma
          Chromosome Location:
          Chr.17: 34122718 - 34139101 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439226_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-DMa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010386.4 NP_034516.3 2 - 3 687 91
          GenBank mRNA AK146950.1 - 2 - 3 688 91
          AK160460.1 - 1 - 2 166 91
          AK169900.1 - 1 - 2 91
          BC001996.1 - 1 - 2 3591389 91
          BC049992.1 - 1 - 2 100 6394795 91
          KJ650223.1 - 1 - 2 91
          U35324.1 - 1 - 2 95 91
          U35325.1 - 1 - 2 91
          U35326.1 - 1 - 2 91
          U35327.1 - 1 - 2 95 91
          X62742.1 - 1 - 2 91
          X90783.1 - 1 - 2 91
          X90784.1 - 1 - 2 91
          X90785.1 - 1 - 2 95 91
          X90786.1 - 1 - 2 95 91
          X90787.1 - 1 - 2 95 91
          X90788.1 - 1 - 2 91
          X90789.1 - 1 - 2 91
          X90790.1 - 1 - 2 95 91
          X90791.1 - 1 - 2 91
          X90792.1 - 1 - 2 91
          X90793.1 - 1 - 2 95 91
          X90794.1 - 1 - 2 95 91
          X90795.1 - 1 - 2 95 91
          X90963.1 - 1 - 2 95 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-DMa

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          peptide antigen assembly with MHC class II protein complex
          antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          positive regulation of germinal center formation
          positive regulation of humoral immune response
          protein complex assembly
          immune response
          protein transport
          immunoglobulin mediated immune response
          antigen processing and presentation
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          positive thymic T cell selection
          positive regulation of T cell differentiation
          inner ear development
          positive regulation of immune response
          chaperone mediated protein folding requiring cofactor
          protein binding
          MHC class II protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90794.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P473734 X90794.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30414 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90794.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P473734 X90794.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90963.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90792.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90793.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90794.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90963.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90792.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90793.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473734 X90794.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152311 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08764661 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08764651 - -
          Whole Mouse Genome G4122A A_51_P473734 X90795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473734 X90794.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115407 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13316652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13316654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13316651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13456794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13518624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13407412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13456815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13099327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13099324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444235 - -
          Affymetrix HT MG-430 PM Array Plate 1422527_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459872_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444229 - -
          Mouse Genome 430 2.0 Array 1459872_x_at - -
          Mouse Genome 430 2.0 Array 1422527_at - -
          Mouse Genome 430 2.0 Array 1422527_PM_at - -
          Mouse Genome 430 2.0 Array 1459872_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14998_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14998_at - -
          Mouse Expression Set 430 1422527_at - -
          Mouse Expression Set 430 1422527_PM_at - -
          Mouse Expression Set 430 1459872_PM_x_at - -
          Mouse Expression Set 430 1459872_x_at - -
          Murine Genome U74v2 Set 93092_PM_at - -
          Murine Genome U74v2 Set 93092_at - -
          Murine Genome U74v2 Set 162346_PM_f_at - -
          Murine Genome U74v2 Set 162346_f_at - -
          Mouse Genome 430A 2.0 Array 1422527_at - -
          Mouse Genome 430A 2.0 Array 1422527_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422527_at - -
          Affymetrix HT Mouse Genome 430A Array 1422527_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459872_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1459872_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 745275 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6290463 - -
          Illumina mouse-6 v1.0 expression beadchip 6290463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2777769 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2777769 - -
          mouse-6 v1.1 (Illumina) scl014998.4_34-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014998.4_34-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2866856 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777769 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2866856 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl014998.4_34-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline