Search
Search

Itgb5| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145884.1 | NP_001139356.1 | 3 - 4 | 660 | 60 | |
| NM_010580.2 | NP_034710.2 | 3 - 4 | 660 | 60 | ||
| GenBank mRNA | AF022110.1 | - | 3 - 4 | 659 | 60 | |
| AF043256.1 | - | 3 - 4 | 565 | 60 | ||
| AF043257.1 | - | 3 - 4 | 565 | 60 | ||
| AK087476.1 | - | 3 - 4 | 406 | 60 | ||
| AK140251.1 | - | 3 - 4 | 572 | 60 | ||
| AK143532.1 | - | 3 - 4 | 595 | 60 | ||
| BC046280.1 | - | 3 - 4 | 569 | 6394641 | 60 | |
| BC058246.1 | - | 3 - 4 | 624 | 6313668 | 60 | |
Set Membership: |
|