Search
Search

Jarid2| Gene Symbol: Jarid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205043.1 | NP_001191972.1 | 10 - 11 | 3253 | 101 | |
| NM_001205044.1 | NP_001191973.1 | 11 - 12 | 2926 | 101 | ||
| NM_021878.3 | NP_068678.1 | 11 - 12 | 2972 | 101 | ||
| GenBank mRNA | AK045214.1 | - | 10 - 11 | 3058 | 101 | |
| AK147226.1 | - | 10 - 11 | 3253 | 101 | ||
| AK164134.1 | - | 9 - 10 | 2577 | 101 | ||
| BC003374.1 | - | 4 - 5 | 822 | 101 | ||
| BC026056.1 | - | 10 - 11 | 3160 | 101 | ||
| BC050129.1 | - | 10 - 11 | 3174 | 101 | ||
| BC051010.1 | - | 10 - 11 | 3156 | 101 | ||
| BC052444.1 | - | 11 - 12 | 2843 | 6406875 | 101 | |
| BC060695.1 | - | 11 - 12 | 2843 | 6406875 | 101 | |
| BC115758.1 | - | 5 - 6 | 1857 | 101 | ||
| D31967.1 | - | 11 - 12 | 2862 | 101 | ||
Set Membership: |
|