Search
Search

Srd5a2| Gene Symbol: Srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053188.2 | NP_444418.1 | 3 - 4 | 633 | 65 | |
| GenBank mRNA | AB049456.1 | - | 3 - 4 | 633 | 65 | |
| AK138946.1 | - | 3 - 4 | 574 | 65 | ||
| BC125510.1 | - | 3 - 4 | 617 | 40130025 | 65 | |
Set Membership: |
|