Search
Search

Neil1| Gene Symbol: Neil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028347.2 | NP_082623.1 | 7 - 8 | 937 | 136 | |
| GenBank mRNA | AB079069.1 | - | 7 - 8 | 937 | 136 | |
| AK013322.1 | - | 8 - 9 | 1008 | 136 | ||
| AK155405.1 | - | 9 - 10 | 813 | 136 | ||
| BC034873.1 | - | 8 - 9 | 1181 | 135 | ||
Set Membership: |
|