Search
Search

Bicd2| Gene Symbol: Bicd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039179.2 | NP_001034268.1 | 5 - 6 | 2219 | 74 | |
| NM_001039180.2 | NP_001034269.1 | 6 - 7 | 2115 | 74 | ||
| NM_029791.4 | NP_084067.1 | 5 - 6 | 2219 | 74 | ||
| GenBank mRNA | AJ250106.1 | - | 5 - 6 | 2178 | 74 | |
| AK122348.1 | - | 5 - 6 | 2192 | 74 | ||
| AK156407.1 | - | 5 - 6 | 2174 | 74 | ||
| AK171300.1 | - | 6 - 7 | 2073 | 74 | ||
| BC032198.1 | - | 5 - 6 | 2176 | 5324113 | 74 | |
Set Membership: |
|