Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460121_m1
          See other INSIG2 GE Assays ›
          Gene Symbol:
          Insig2
          Gene Name:
          insulin induced gene 2
          Gene Aliases:
          2900053I11Rik, C730043J18Rik, Insig-2
          Chromosome Location:
          Chr.1: 121304353 - 121333654 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460121_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Insig2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271531.1 NP_001258460.1 3 - 4 591 80
          NM_001271532.1 NP_001258461.1 2 - 3 458 80
          NM_133748.2 NP_598509.1 3 - 4 834 80
          NM_178082.3 NP_835183.1 3 - 4 598 80
          GenBank mRNA AF527631.1 - 3 - 4 568 80
          AK040703.1 - 3 - 4 589 80
          AK050394.1 - 3 - 4 592 80
          AK152076.1 - 3 - 4 564 80
          AK154543.1 - 3 - 4 597 80
          AK161256.1 - 3 - 4 602 80
          AK165634.1 - 3 - 4 603 80
          AK190310.1 - 1 - 2 69 80
          BC015288.1 - 3 - 4 548 4240711 80
          BC023067.1 - 3 - 4 572 5357599 80
          BC023874.1 - 2 - 3 490 5355039 80
          BC024411.1 - 3 - 4 586 4971297 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002969.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Insig2

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          triglyceride metabolic process
          cholesterol biosynthetic process
          response to sterol depletion
          steroid metabolic process
          cholesterol metabolic process
          negative regulation of steroid biosynthetic process
          sterol biosynthetic process
          SREBP signaling pathway
          inner ear morphogenesis
          middle ear morphogenesis
          negative regulation of fatty acid biosynthetic process
          palate development
          cranial suture morphogenesis
          protein binding
          transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100023913.mm.1 NM_001271531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023913.mm.1 NM_001271532.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023913.mm.1 NM_133748.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023913.mm.1 NM_178082.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023913.mm.1 TC0100002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023914.mm.1 NM_001271531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023914.mm.1 NM_001271532.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023914.mm.1 NM_133748.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023914.mm.1 NM_178082.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023914.mm.1 TC0100002969.mm.1 MTA TC

          Back To Top
           

          Citations (1)

          Complex role of the vitamin D receptor and its ligand in adipogenesis in 3T3-L1 cells.
          Authors
          Blumberg JM, Tzameli I, Astapova I, Lam FS, Flier JS, Hollenberg AN
          Journal
          J Biol Chem - Page No 11205 (Fri Apr 21 00:00:00 UTC 2006)
          Volume (Year)
          281 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline