Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460981_g1
          See other GPS1 GE Assays ›
          Gene Symbol:
          Gps1
          Gene Name:
          G protein pathway suppressor 1
          Gene Aliases:
          Cops1, Csn1, R75577, Sgn1
          Chromosome Location:
          Chr.11: 120784260 - 120789107 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460981_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gps1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145370.2 NP_663345.2 1 - 2 214 66
          GenBank mRNA BC003350.1 - 1 - 2 214 3481979 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002048.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC003350.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gps1

          Gene Ontology Categories:

          Function(s) Process(es)

          cullin deneddylation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186730 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186732 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087385 BC103782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087385 AK088490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087385 BC003350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087385 NM_145370.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087385 NM_001177874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087384 BC103782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087384 BC003350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087384 NM_145370.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087384 NM_001177874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087384 AK049333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52058 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149786 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087385 BC103782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087385 AK088490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087385 BC003350.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087385 NM_145370.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087385 NM_001177874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143968 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087384 BC103782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087384 BC003350.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087384 NM_145370.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087384 NM_001177874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087384 AK049333.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186754 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149786 - -
          Whole Mouse Genome G4122A A_51_P374453 BC103782.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374453 AK088490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374453 BC003350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374453 NM_145370.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374453 NM_001177874.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114780 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121771 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149786 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09736791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09736770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09736783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09736789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09736816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09736772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09736815 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10383500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383486 - -
          Affymetrix HT MG-430 PM Array Plate 1415699_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436446 - -
          Affymetrix Mouse Gene 1.1 ST Array 10383485 - -
          Mouse Genome 430 2.0 Array 1415699_a_at - -
          Mouse Genome 430 2.0 Array 1415699_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1100019774.mm.1 NM_145370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019774.mm.1 NM_145370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019774.mm.1 TC1100002048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019773.mm.1 NM_145370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019773.mm.1 NM_145370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019773.mm.1 TC1100002048.mm.1 MTA TC
          Mouse Expression Set 430 1415699_a_at - -
          Mouse Expression Set 430 1415699_PM_a_at - -
          Mouse Expression Set 430 1415699_PM_a_at - -
          Mouse Expression Set 430 1415699_a_at - -
          Murine Genome U74v2 Set 96943_PM_at - -
          Murine Genome U74v2 Set 96943_at - -
          Mouse Genome 430A 2.0 Array 1415699_a_at - -
          Mouse Genome 430A 2.0 Array 1415699_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415699_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415699_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415699_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415699_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 901711 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470037 - -
          Illumina mouse-6 v1.0 expression beadchip 2470037 - -
          Illumina mouse-6 v1.0 expression beadchip 4760315 - -
          Illumina mouse-6 v1.0 expression beadchip 7100040 - -
          Illumina mouse-6 v1.0 expression beadchip 6200528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006200528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254788 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632714 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601090 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625982 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601090 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2632714 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254788 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625982 - -
          mouse-6 v1.1 (Illumina) scl00209318.1_9-S - -
          mouse-6 v1.1 (Illumina) scl0209318.14_24-S - -
          mouse-6 v1.1 (Illumina) scl0209318.1_47-S - -
          mouse-6 v1.1 (Illumina) scl0001480.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760315 - -
          Illumina Sentrix mouse-6.v1 bead array 2470037 - -
          Illumina Sentrix mouse-6.v1 bead array 6200528 - -
          Illumina Sentrix mouse-6.v1 bead array 7100040 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0209318.1_47-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0209318.14_24-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001480.1_10-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00209318.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760315 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470037 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625982 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601090 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2632714 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254788 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254788 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625982 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601090 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2632714 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0209318.14_24-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline