Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466014_m1
          See other AKIRIN2 GE Assays ›
          Gene Symbol:
          Akirin2
          Gene Name:
          akirin 2
          Gene Aliases:
          2700059D21Rik, AA114675, AA522011, AU019887, Akirin-2
          Chromosome Location:
          Chr.4: 34550615 - 34566911 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466014_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akirin2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007589.3 NP_001007590.2 1 - 2 659 88
          GenBank mRNA AK005836.1 - 1 - 2 338 88
          BC082305.1 - 2 - 3 347 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000304.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akirin2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          immune system process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          positive regulation of cell proliferation
          embryo development
          negative regulation of gene expression
          positive regulation of endopeptidase activity
          response to lipopolysaccharide
          positive regulation of interleukin-6 production
          innate immune response
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          protein binding
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831245 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831250 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011967 BC082305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011967 NM_001007589.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011967 AK004133.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41655 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157288 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011967 BC082305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011967 NM_001007589.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011967 AK004133.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22831245 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157288 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00847129 - -
          Whole Mouse Genome G4122A A_52_P442779 BC082305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P442779 NM_001007589.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P442779 AK004133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P442779 AK005836.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157288 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03634570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03634575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03804942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03634569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03804943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03634573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831250 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831245 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22831248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22831346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03934696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03665711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03555826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03612968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03442205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03262433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03804943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03634576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03555828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04027419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03262435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03324978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03797623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22831263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10503924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503921 - -
          Affymetrix HT MG-430 PM Array Plate 1428238_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428237_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10503917 - -
          Mouse Genome 430 2.0 Array 1428238_at - -
          Mouse Genome 430 2.0 Array 1428237_at - -
          Mouse Genome 430 2.0 Array 1428237_PM_at - -
          Mouse Genome 430 2.0 Array 1428238_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433693_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433693_at - -
          Affymetrix Mouse Clariom S Array PSR0400001920.mm.1 NM_001007589.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001920.mm.1 NM_001007589.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001920.mm.1 TC0400000304.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400001921.mm.1 NM_001007589.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001921.mm.1 NM_001007589.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001921.mm.1 TC0400000304.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400001918.mm.1 NM_001007589.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001918.mm.1 NM_001007589.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001918.mm.1 TC0400000304.mm.1 MTA TC
          Mouse Expression Set 430 1428237_PM_at - -
          Mouse Expression Set 430 1428237_at - -
          Mouse Expression Set 430 1428238_PM_at - -
          Mouse Expression Set 430 1428238_at - -
          Murine Genome U74v2 Set 93768_PM_f_at - -
          Murine Genome U74v2 Set 93768_f_at - -
          Murine Genome U74v2 Set 93767_PM_i_at - -
          Murine Genome U74v2 Set 93767_i_at - -
          Mouse Genome 430A 2.0 Array 1428237_PM_at - -
          Mouse Genome 430A 2.0 Array 1428237_at - -
          Mouse Genome 430A 2.0 Array 1428238_at - -
          Mouse Genome 430A 2.0 Array 1428238_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428238_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428238_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 361714 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230192 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline