Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470108_m1
          See other ZDHHC16 GE Assays ›
          Gene Symbol:
          Zdhhc16
          Gene Name:
          zinc finger, DHHC domain containing 16
          Gene Aliases:
          1500015N03Rik, Aph2
          Chromosome Location:
          Chr.19: 41933472 - 41944104 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470108_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zdhhc16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023740.2 NP_076229.2 10 - 11 1315 110
          GenBank mRNA AF176814.1 - 10 - 11 1288 110
          AK005255.1 - 9 - 10 1047 110
          AK151505.1 - 10 - 11 1297 110
          AK168042.1 - 10 - 11 1290 110
          BC010835.1 - 10 - 11 1279 4239913 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000602.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zdhhc16

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          protein palmitoylation
          zinc ion binding
          palmitoyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          protein-cysteine S-palmitoyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117997 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453519 AF176814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453519 AF176814.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6986 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155244 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453519 AF176814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453519 AF176814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P453519 AF176814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P453519 AF176814.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32118002 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03598838 - -
          Whole Mouse Genome G4122A A_51_P453519 AF176814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453519 AF176814.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127907 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126944 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155244 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14374904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14374905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14374893 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32118004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14447801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14412088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14374893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14357106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14374910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14357108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14370726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14374905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14374904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14357102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32118049 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463178 - -
          Affymetrix HT MG-430 PM Array Plate 1421115_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10463173 - -
          Affymetrix Mouse Gene 1.1 ST Array 10467697 - -
          Mouse Genome 430 2.0 Array 1421115_a_at - -
          Mouse Genome 430 2.0 Array 1458646_at - -
          Mouse Genome 430 2.0 Array 1458646_PM_at - -
          Mouse Genome 430 2.0 Array 1421115_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74168_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74168_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900005394.mm.1 NM_023740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005394.mm.1 NM_023740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005394.mm.1 TC1900000602.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005392.mm.1 NM_023740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005392.mm.1 NM_023740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005392.mm.1 TC1900000602.mm.1 MTA TC
          Mouse Expression Set 430 1421115_PM_a_at - -
          Mouse Expression Set 430 1421115_a_at - -
          Murine Genome U74v2 Set 103643_PM_at - -
          Murine Genome U74v2 Set 103643_at - -
          Murine Genome U74v2 Set 170666_PM_i_at - -
          Murine Genome U74v2 Set 170666_i_at - -
          Mouse Genome 430A 2.0 Array 1421115_a_at - -
          Mouse Genome 430A 2.0 Array 1421115_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421115_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421115_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 573205 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6180324 - -
          Illumina mouse-6 v1.0 expression beadchip 6180324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2493874 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2452849 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493874 - -
          mouse-6 v1.1 (Illumina) scl53123.9.6_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53123.9.6_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2493874 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2964608 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2493874 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2964608 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53123.9.6_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline