Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473193_m1
          See other PRAMEL1 GE Assays ›
          Gene Symbol:
          Pramel1
          Gene Name:
          preferentially expressed antigen in melanoma-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 143394359 - 143400160 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473193_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pramel1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031377.2 NP_113554.1 3 - 4 986 123
          GenBank mRNA AF285578.1 - 3 - 4 1021 123
          BC131995.1 - 3 - 4 987 40130272 123
          BC132399.1 - 3 - 4 987 40130676 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001824.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pramel1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374912 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374915 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374913 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374911 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374916 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026465 AF285578.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39431 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23374909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154536 - -
          Whole Mouse Genome G4122A A_51_P174783 BC131995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174783 BC132399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174783 NM_031377.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174783 AF285578.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134395 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109948 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03437817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03437818 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374915 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374913 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374911 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374916 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23374912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23375017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23374909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03994502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03378930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23374956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03994506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03499126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510059 - -
          Affymetrix HT MG-430 PM Array Plate 1421713_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10510056 - -
          Mouse Genome 430 2.0 Array 1421713_at - -
          Mouse Genome 430 2.0 Array 1421713_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83491_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83491_at - -
          Affymetrix Mouse Clariom S Array PSR0400015052.mm.1 NM_031377.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015052.mm.1 NM_031377.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015052.mm.1 TC0400001824.mm.1 MTA TC
          Mouse Expression Set 430 1421713_at - -
          Mouse Expression Set 430 1421713_PM_at - -
          Mouse Genome 430A 2.0 Array 1421713_at - -
          Mouse Genome 430A 2.0 Array 1421713_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421713_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421713_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 381011 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4480014 - -
          Illumina mouse-6 v1.0 expression beadchip 4480014 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480014 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253686 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253686 - -
          mouse-6 v1.1 (Illumina) scl24730.4.1_103-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480014 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24730.4.1_103-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480014 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1253686 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253686 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline