Search
Search

Ap1m1| Gene Symbol: Ap1m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007456.4 | NP_031482.1 | 3 - 4 | 335 | 72 | |
| GenBank mRNA | AK148173.1 | - | 3 - 4 | 350 | 72 | |
| AK215581.1 | - | 3 - 4 | 245 | 72 | ||
| BC003823.1 | - | 3 - 4 | 309 | 3491338 | 72 | |
| M62419.1 | - | 3 - 4 | 337 | 72 | ||
Set Membership: |
|