Search
Search
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007482.3 | NP_031508.1 | 4 - 5 | 573 | 75 | |
| GenBank mRNA | AK149471.1 | - | 4 - 5 | 556 | 75 | |
| AK171417.1 | - | 3 - 4 | 407 | 75 | ||
| BC013341.1 | - | 4 - 5 | 550 | 4163626 | 75 | |
| BC050005.1 | - | 4 - 5 | 587 | 3990246 | 75 | |
| CB947842.1 | - | 4 - 5 | 577 | 75 | ||
| CT010173.1 | - | 4 - 5 | 471 | 75 | ||
| U51805.1 | - | 4 - 5 | 612 | 75 | ||
Set Membership: |
|