| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124016 |
NM_008698.2 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654643 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726025 |
AB030187.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726025 |
AK013203.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726025 |
AK018154.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726025 |
AK163183.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726025 |
BC027164.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726025 |
NM_022424.5 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299582 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
870846 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
424479 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
76745 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
719241 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
488187 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
269625 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
711153 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689561 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658998 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198567 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
37226 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
57277 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149012 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110702 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124016 |
NM_008698.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P214516 |
AJ001260.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P214516 |
NM_008698.2 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560095 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560086 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560165 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560075 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560185 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560080 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560182 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560083 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560085 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560159 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560087 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560079 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560163 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560168 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560076 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560093 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560179 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560176 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560169 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560162 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560073 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560167 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560172 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560171 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560161 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560184 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560166 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560157 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560177 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560189 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560096 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560181 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560091 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560089 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27560186 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07241858 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110702 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149012 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07241873 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P214516 |
AJ001260.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P214516 |
NM_008698.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117105 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149012 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110702 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09516251 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09516249 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09177186 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09516254 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09516250 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560182 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560169 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560168 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560122 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560071 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560235 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560176 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560089 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560085 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560177 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560109 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560130 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560117 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560239 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560113 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560115 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560149 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27560066 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560179 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560163 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560105 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09399831 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09489194 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560082 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09516249 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560080 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560067 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560176 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09249260 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560083 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560140 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09399829 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560159 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560081 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560103 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560172 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560066 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09399833 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560101 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09155180 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560186 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560100 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560161 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09352214 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560077 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560167 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560164 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560169 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09226074 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560182 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09315477 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560171 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560148 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560150 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560075 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560106 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560091 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560142 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560093 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560094 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560173 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560147 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560099 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09177185 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09226064 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560157 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560087 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560189 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560166 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09721473 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560165 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09399820 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560070 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560076 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09584473 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560149 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560074 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560097 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560177 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560064 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09723515 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27560096 |
- |
- |