Search
Search

Gabarap| Gene Symbol: Gabarap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019749.4 | NP_062723.1 | 2 - 3 | 514 | 90 | |
| GenBank mRNA | AF161587.1 | - | 2 - 3 | 234 | 90 | |
| AK002879.1 | - | 2 - 3 | 269 | 90 | ||
| AK011731.1 | - | 2 - 3 | 179 | 90 | ||
| AV029091.2 | - | 2 - 3 | 285 | 90 | ||
| BC002126.1 | - | 2 - 3 | 277 | 3497484 | 90 | |
| BC024621.1 | - | 2 - 3 | 297 | 4459651 | 90 | |
| BC029329.1 | - | 2 - 3 | 216 | 90 | ||
| BC030350.1 | - | 2 - 3 | 523 | 5102114 | 90 | |
Set Membership: |
|