Search
Search

Cenpk| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021790.2 | NP_068562.1 | 2 - 3 | 422 | 124 | |
| NM_181061.5 | NP_851406.1 | 2 - 3 | 545 | 124 | ||
| NR_075088.1 | 2 - 3 | 545 | 124 | |||
| GenBank mRNA | AB043687.1 | - | 2 - 3 | 422 | 124 | |
| AK082939.1 | - | 2 - 3 | 202 | 124 | ||
| AK160776.1 | - | 2 - 3 | 180 | 124 | ||
| BC028655.1 | - | 2 - 3 | 545 | 124 | ||
| BC092351.1 | - | 2 - 3 | 184 | 124 | ||
Set Membership: |
|