Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491491_m1
          See other CAPN12 GE Assays ›
          Gene Symbol:
          Capn12
          Gene Name:
          calpain 12
          Gene Aliases:
          CANP 12
          Chromosome Location:
          Chr.7: 28881446 - 28893585 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491491_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capn12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110807.1 NP_001104277.1 4 - 5 629 63
          GenBank mRNA AJ289243.1 - 4 - 5 637 63
          BC028751.1 - 4 - 5 628 1314413 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000517.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          calcium-dependent cysteine-type endopeptidase activity
          calcium ion binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 AK145322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 AJ289242.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 NM_021895.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 AK029042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 BC028751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 AK145322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 AJ289242.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 NM_021895.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 AK029042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644264 BC028751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224282 BC028751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224282 NM_001110807.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224282 AJ289243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224282 BC028751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224282 NM_001110807.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224282 AJ289243.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130496 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 AK145322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 AJ289242.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 NM_021895.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 AK029042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 BC028751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 AK145322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 AJ289242.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 NM_021895.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 AK029042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644264 BC028751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224282 BC028751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224282 NM_001110807.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224282 AJ289243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224282 BC028751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224282 NM_001110807.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224282 AJ289243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 AK145322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 AJ289242.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 NM_021895.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 AK029042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 BC028751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 AK145322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 AJ289242.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 NM_021895.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 AK029042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644264 BC028751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224282 BC028751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224282 NM_001110807.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224282 AJ289243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224282 BC028751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224282 NM_001110807.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224282 AJ289243.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05901217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06133558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06520925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06412076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06085705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06647257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06520927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05901230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05901242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05901235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06140994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25024076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551652 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551655 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551651 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551658 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551653 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551650 - -
          Affymetrix HT MG-430 PM Array Plate 1449970_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551644 - -
          Affymetrix Mouse Gene 1.1 ST Array 10561527 - -
          Mouse Genome 430 2.0 Array 1444258_PM_at - -
          Mouse Genome 430 2.0 Array 1444258_at - -
          Mouse Genome 430 2.0 Array 1449970_at - -
          Mouse Genome 430 2.0 Array 1449970_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60594_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60594_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700004339.mm.1 NM_001110807.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004339.mm.1 NM_001110807.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004339.mm.1 TC0700000517.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004340.mm.1 NM_001110807.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004340.mm.1 NM_001110807.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004340.mm.1 TC0700000517.mm.1 MTA TC
          Mouse Expression Set 430 1449970_PM_at - -
          Mouse Expression Set 430 1449970_at - -
          Mouse Genome 430A 2.0 Array 1449970_at - -
          Mouse Genome 430A 2.0 Array 1449970_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449970_at - -
          Affymetrix HT Mouse Genome 430A Array 1449970_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763302 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2100050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666371 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666371 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2666371 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666371 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline