Search
Search

Trim28| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 1 - 2 | 909 | 83 | |
| GenBank mRNA | AF230391.1 | - | 1 - 2 | 510 | 83 | |
| AF230392.1 | - | 1 - 2 | 510 | 83 | ||
| AK141357.1 | - | 1 - 2 | 653 | 83 | ||
| BC020452.1 | - | 1 - 2 | 340 | 83 | ||
| BC023335.1 | - | 1 - 2 | 618 | 83 | ||
| BC058391.1 | - | 1 - 2 | 355 | 6817165 | 83 | |
| BC089337.1 | - | 1 - 2 | 909 | 6811294 | 83 | |
| U67303.1 | - | 1 - 2 | 900 | 83 | ||
| X99644.1 | - | 1 - 2 | 510 | 83 | ||
Set Membership: |
|