Search
Search

Actn3| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013456.2 | NP_038484.1 | 17 - 18 | 2231 | 72 | |
| GenBank mRNA | AF093775.1 | - | 17 - 18 | 2206 | 72 | |
| AK134757.1 | - | 2 - 3 | 335 | 72 | ||
| BC111890.1 | - | 17 - 18 | 2197 | 40060598 | 72 | |
Set Membership: |
|