Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00500348_m1
          See other CAPG GE Assays ›
          Gene Symbol:
          Capg
          Gene Name:
          capping protein (actin filament), gelsolin-like
          Gene Aliases:
          gCap39, mbh1
          Chromosome Location:
          Chr.6: 72544391 - 72562983 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00500348_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042534.3 NP_001035999.1 9 - 10 1233 100
          NM_001271395.1 NP_001258324.1 9 - 10 1226 100
          NM_001271415.1 NP_001258344.1 10 - 11 1223 100
          NM_007599.4 NP_031625.2 9 - 10 1080 100
          GenBank mRNA AK146622.1 - 9 - 10 1084 100
          AK152818.1 - 9 - 10 1058 100
          AK159577.1 - 9 - 10 1033 100
          AK159736.1 - 9 - 10 1035 100
          AK165153.1 - 9 - 10 1047 100
          AK165586.1 - 7 - 8 876 100
          BC023101.1 - 9 - 10 1022 3983574 100
          X54511.1 - 10 - 11 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000852.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capg

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell projection assembly
          barbed-end actin filament capping
          actin filament capping
          actin binding
          protein domain specific binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998631 AK159577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998631 AK159736.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998631 AK165153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998631 AK165586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998631 NM_007599.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85599 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119171 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144637 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998631 AK159577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998631 AK159736.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998631 AK165153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998631 AK165586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998631 NM_007599.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273921 X54511.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24542217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144637 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119171 - -
          Whole Mouse Genome G4122A A_51_P273921 X54511.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116893 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144637 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05085959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05085944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24542236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05713710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05442048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05085944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05758116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05654672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05276157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05877942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05656844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05656835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05085953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05429376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05218846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05599094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05656846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05135226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05085956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05085959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05656833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05138850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05877938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05085955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05276156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05747876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05344099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24542188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05656834 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539139 - -
          Affymetrix HT MG-430 PM Array Plate 1450355_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1447803_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539135 - -
          Mouse Genome 430 2.0 Array 1450355_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450355_a_at - -
          Mouse Genome 430 2.0 Array 1447803_PM_x_at - -
          Mouse Genome 430 2.0 Array 1447803_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12332_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12332_at - -
          Affymetrix Mouse Clariom S Array PSR0600006553.mm.1 NM_001042534.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006553.mm.1 NM_001271395.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006553.mm.1 NM_001271415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006553.mm.1 NM_007599.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006553.mm.1 TC0600000852.mm.1 MTA TC
          Mouse Expression Set 430 1450355_a_at - -
          Mouse Expression Set 430 1450355_PM_a_at - -
          Mouse Expression Set 430 1447803_x_at - -
          Mouse Expression Set 430 1447803_PM_x_at - -
          Murine Genome U74v2 Set 161969_PM_f_at - -
          Murine Genome U74v2 Set 162461_PM_at - -
          Murine Genome U74v2 Set 162461_at - -
          Murine Genome U74v2 Set 160106_at - -
          Murine Genome U74v2 Set 160106_PM_at - -
          Murine Genome U74v2 Set 161969_f_at - -
          Murine Genome U74v2 Set 166798_PM_at - -
          Murine Genome U74v2 Set 166798_at - -
          Mouse Genome 430A 2.0 Array 1450355_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450355_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450355_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450355_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1447803_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447803_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770373 - -
          Illumina mouse-6 v1.0 expression beadchip 770373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773113 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773113 - -
          mouse-6 v1.1 (Illumina) scl29881.14.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 770373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29881.14.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770373 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2773113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3115281 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3115281 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2773113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29881.14.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline