Search
Search

Pcsk2| Gene Symbol: Pcsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008792.4 | NP_032818.1 | 3 - 4 | 749 | 110 | |
| GenBank mRNA | AF483512.1 | - | 3 - 4 | 391 | 110 | |
| AF483513.1 | - | 3 - 4 | 391 | 110 | ||
| AK133620.1 | - | 3 - 4 | 750 | 110 | ||
| AK158493.1 | - | 3 - 4 | 697 | 110 | ||
| AK158598.1 | - | 3 - 4 | 811 | 110 | ||
| AK161585.1 | - | 3 - 4 | 690 | 110 | ||
| BC052013.1 | - | 3 - 4 | 666 | 5707923 | 110 | |
| BC057348.1 | - | 3 - 4 | 704 | 6827695 | 110 | |
| M55669.1 | - | 3 - 4 | 475 | 110 | ||
Set Membership: |
|