Search
Search
| Gene Symbol: Mlh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029786.1 | - | 4 - 5 | 2137 | 95 | |
| AK132195.1 | - | 4 - 5 | 3545 | 95 | ||
| AK162464.1 | - | 3 - 4 | 2212 | 95 | ||
| BC003865.1 | - | 3 - 4 | 339 | 95 | ||
| BC072599.1 | - | 4 - 5 | 3556 | 95 | ||
| BC079861.1 | - | 4 - 5 | 3543 | 6834767 | 95 | |
Set Membership: |
|