Search
Search

Wasf3| Gene Symbol: Wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145155.3 | NP_660137.1 | 6 - 7 | 964 | 64 | |
| GenBank mRNA | AF454703.1 | - | 5 - 6 | 583 | 64 | |
| AK044481.1 | - | 985 | 64 | |||
| BC027038.1 | - | 6 - 7 | 964 | 5363785 | 64 | |
Set Membership: |
|