Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528168_s1
          See other OLFR640 GE Assays ›
          Gene Symbol:
          Olfr640
          Gene Name:
          olfactory receptor 640
          Gene Aliases:
          MOR13-4
          Chromosome Location:
          Chr.7: 104021372 - 104022316 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528168_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr640
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146822.2 NP_667033.2 1 54 77
          GenBank mRNA BC106802.2 - 1 54 40042505 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003922.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr640

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046223 NM_146822.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046223 BC106802.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657508 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046223 NM_146822.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046223 BC106802.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P101022 NM_146822.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P101022 BC106802.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386249 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06308159 - -
          Whole Mouse Genome G4122A A_51_P101022 NM_146822.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P101022 BC106802.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112222 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06308159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566295 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566294 - -
          Affymetrix Mouse Clariom S Array PSR0700032833.mm.1 NM_146822.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032833.mm.1 NM_146822.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032833.mm.1 TC0700003922.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 826367 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6100427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100427 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644084 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644084 - -
          mouse-6 v1.1 (Illumina) scl8864.1.1_123-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8864.1.1_123-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2644084 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2644084 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline