Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558700_m1
          See other TNFRSF26 GE Assays ›
          Gene Symbol:
          Tnfrsf26
          Gene Name:
          tumor necrosis factor receptor superfamily, member 26
          Gene Aliases:
          Tnfrh3, Tnfrsf24
          Chromosome Location:
          Chr.7: 143607685 - 143627845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558700_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175649.5 NP_783580.1 4 - 5 382 69
          GenBank mRNA AK142511.1 - 443 69
          AK154666.1 - 4 - 5 493 69
          AY165628.1 - 4 - 5 382 69
          BC111108.1 - 4 - 5 370 69
          BC138819.1 - 4 - 5 431 8861841 69
          BC138820.1 - 4 - 5 432 8861842 69
          BC152357.1 - 4 - 5 378 69
          BC152358.1 - 4 - 5 382 40055748 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004591.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf26

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 BC138819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 BC138820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 AK154666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 AY165628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 NM_175649.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 BC138819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 BC138820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 AK154666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 AY165628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554536 NM_175649.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126952 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148432 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172971 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 BC138819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 BC138820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 AK154666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 AY165628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 NM_175649.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 BC138819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 BC138820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 AK154666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 AY165628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554536 NM_175649.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172971 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148432 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01659590 - -
          Whole Mouse Genome G4122A A_52_P554536 BC138819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 BC138820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 AK154666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 AY165628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 NM_175649.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 BC138819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 BC138820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 AK154666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 AY165628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554536 NM_175649.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108004 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172971 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06150057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06580626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06400946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06400945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06144801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05983901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06153117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06533491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06306871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06474571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05887713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06001968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602102 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569490 - -
          Affymetrix Mouse Gene 1.1 ST Array 10569485 - -
          Affymetrix Mouse Clariom S Array PSR0700038810.mm.1 NM_175649.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038810.mm.1 NM_175649.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038810.mm.1 TC0700004591.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 736058 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2479124 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2479124 - -
          mouse-6 v1.1 (Illumina) scl0244237.1_299-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0244237.1_299-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2479124 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2479124 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline