Search
Search
| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178378.3 | NP_848465.1 | 4 - 5 | 554 | 79 | |
| GenBank mRNA | AK146422.1 | - | 2 - 3 | 292 | 79 | |
| BC049990.1 | - | 4 - 5 | 538 | 6310844 | 79 | |
Set Membership: |
|