Search
Search

Ccbl2| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 8 - 9 | 925 | 104 | |
| NM_173763.4 | NP_776124.1 | 8 - 9 | 929 | 104 | ||
| GenBank mRNA | AF363737.1 | - | 8 - 9 | 899 | 104 | |
| AK049569.1 | - | 8 - 9 | 898 | 104 | ||
| AK145623.1 | - | 8 - 9 | 923 | 104 | ||
| BC037135.1 | - | 4 - 5 | 506 | 104 | ||
| BC085287.1 | - | 9 - 10 | 1022 | 104 | ||
| BC131942.1 | - | 7 - 8 | 714 | 40130219 | 104 | |
| BC132615.1 | - | 7 - 8 | 714 | 40130892 | 104 | |
Set Membership: |
|