Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622644_s1
          See other LRRC31 GE Assays ›
          Gene Symbol:
          Lrrc31
          Gene Name:
          leucine rich repeat containing 31
          Gene Aliases:
          E230002P03Rik
          Chromosome Location:
          Chr.3: 30678917 - 30699840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00622644_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc31
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK053932.1 - 1 1445 101
          AK085279.1 - 2 504 101

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078994 AK136409.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123615 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154449 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064701 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078994 AK136409.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064702 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04861681 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123615 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154449 - -
          Whole Mouse Genome G4122A A_52_P363013 AK053932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363013 AK085279.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123615 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02956480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02475117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03149831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03177721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02672841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02460093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02460089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03045304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02364287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03199676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02430918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03177722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03177730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03149830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02670880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02572902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22029715 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1447173_PM_at - -
          Mouse Genome 430 2.0 Array 1447173_at - -
          Mouse Genome 430 2.0 Array 1447173_PM_at - -
          Mouse Expression Set 430 1447173_at - -
          Mouse Expression Set 430 1447173_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 593317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline