Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625293_m1
          See other KLHL23 GE Assays ›
          Gene Symbol:
          Klhl23
          Gene Name:
          kelch-like 23
          Gene Aliases:
          C130068N17Rik
          Chromosome Location:
          Chr.2: 69821920 - 69836651 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625293_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klhl23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177784.4 NP_808452.2 3 - 4 1637 93
          GenBank mRNA AK048518.1 - 3 - 4 1536 93
          BC072626.1 - 3 - 4 1637 6827841 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001012.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klhl23

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311220 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994922 NM_177784.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994922 AI425639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994922 AK048518.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140712 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115888 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994922 NM_177784.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994922 AI425639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994922 AK048518.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21311200 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115888 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04557011 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04557040 - -
          Whole Mouse Genome G4122A A_52_P289559 NM_177784.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289559 AK048518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289559 BC072626.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459787 NM_177784.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459787 AK048518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517508 NM_177784.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517508 AK048518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517508 BC072626.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117858 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02156199 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02044985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02156205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21311218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21311331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01262272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01915097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01483224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01972752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01517349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01300117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01445054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01413448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02146758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01338945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01999096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02031071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01445052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01548554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01555722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01475645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21311263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02156199 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10472601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472602 - -
          Affymetrix HT MG-430 PM Array Plate 1435743_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10472598 - -
          Mouse Genome 430 2.0 Array 1435743_PM_at - -
          Mouse Genome 430 2.0 Array 1435743_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 277396_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 277396_at - -
          Affymetrix Mouse Clariom S Array PSR0200007996.mm.1 NM_177784.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007996.mm.1 NM_177784.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007996.mm.1 TC0200001012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007997.mm.1 NM_177784.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007997.mm.1 NM_177784.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007997.mm.1 TC0200001012.mm.1 MTA TC
          Mouse Expression Set 430 1435743_PM_at - -
          Mouse Expression Set 430 1435743_at - -
          Murine Genome U74v2 Set 171322_PM_at - -
          Murine Genome U74v2 Set 171322_at - -
          Murine Genome U74v2 Set 168297_PM_f_at - -
          Murine Genome U74v2 Set 168297_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1435743_at - -
          Affymetrix HT Mouse Genome 430B Array 1435743_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550004 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601195 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601195 - -
          mouse-6 v1.1 (Illumina) scl0277396.4_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0277396.4_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862875 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862875 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline