Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00656868_g1
          See other LHB GE Assays ›
          Gene Symbol:
          Lhb
          Gene Name:
          luteinizing hormone beta
          Gene Aliases:
          LH, LH-B, LSH-B, LSH-beta
          Chromosome Location:
          Chr.7: 45418166 - 45421855 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00656868_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lhb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008497.2 NP_032523.2 2 - 3 189 134
          GenBank mRNA AV173269.2 - 1 - 2 201 134
          BB618381.1 - 2 - 3 190 134
          BM224915.2 - 1 - 2 134
          Y10418.1 - 1 - 2 133 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000846.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lhb

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cGMP metabolic process
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953991 NM_008497.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953991 BB618381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953991 BM224915.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953991 Y10418.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242712 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953991 NM_008497.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953991 BB618381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953991 BM224915.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953991 Y10418.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476529 NM_008497.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476529 AV173269.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476529 BB618381.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476529 BM224915.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476529 Y10418.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093029 - -
          Whole Mouse Genome G4122A A_52_P658479 - -
          Whole Mouse Genome G4122A A_51_P476529 NM_008497.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476529 AV173269.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476529 BB618381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476529 BM224915.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476529 Y10418.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128875 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107643 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06205013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06204972 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06205001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06204992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06205014 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06204967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06205000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06205006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06205014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552944 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552943 - -
          Affymetrix HT MG-430 PM Array Plate 1450795_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552942 - -
          Mouse Genome 430 2.0 Array 1450795_PM_at - -
          Mouse Genome 430 2.0 Array 1450795_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16866_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16866_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700007274.mm.1 NM_008497.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007274.mm.1 NM_008497.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007274.mm.1 TC0700000846.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007275.mm.1 NM_008497.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007275.mm.1 NM_008497.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007275.mm.1 TC0700000846.mm.1 MTA TC
          Mouse Expression Set 430 1450795_at - -
          Mouse Expression Set 430 1450795_PM_at - -
          Murine Genome U74v2 Set 101738_at - -
          Murine Genome U74v2 Set 101738_PM_at - -
          Mouse Genome 430A 2.0 Array 1450795_at - -
          Mouse Genome 430A 2.0 Array 1450795_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450795_at - -
          Affymetrix HT Mouse Genome 430A Array 1450795_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105900463 - -
          Illumina mouse-6 v1.0 expression beadchip 105900463 - -
          Illumina mouse-6 v1.0 expression beadchip 1450368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589011 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248017 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589011 - -
          mouse-6 v1.1 (Illumina) scl0016866.2_28-S - -
          mouse-6 v1.1 (Illumina) scl32684.3.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 105900463 - -
          Illumina Sentrix mouse-6.v1 bead array 1450368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016866.2_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450368 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105900463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589011 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline