Search
Search
| Gene Symbol: Lsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017405.2 | NP_059101.1 | 4 - 5 | 919 | 98 | |
| GenBank mRNA | BC004672.1 | - | 4 - 5 | 665 | 3500090 | 98 |
| U49507.1 | - | 4 - 5 | 810 | 98 | ||
Set Membership: |
|