Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663036_m1
          See other TCTA GE Assays ›
          Gene Symbol:
          Tcta
          Gene Name:
          T cell leukemia translocation altered gene
          Gene Aliases:
          9130410M22Rik, AW553637, C85065, Tctal
          Chromosome Location:
          Chr.9: 108302958 - 108306125 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663036_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcta
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133986.1 NP_598747.1 2 - 3 333 62
          GenBank mRNA AK133663.1 - 372 62
          BC019397.1 - 2 - 3 333 3672301 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003058.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcta

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26814196 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26814194 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26814197 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P280401 AK133663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P280401 BC019397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P280401 NM_133986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P280401 AK133663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P280401 BC019397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P280401 NM_133986.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408136 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179948 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168109 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P280401 AK133663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P280401 BC019397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P280401 NM_133986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P280401 AK133663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P280401 BC019397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P280401 NM_133986.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814195 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179948 - -
          Whole Mouse Genome G4122A A_51_P280401 AK133663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280401 BC019397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280401 NM_133986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280401 AK133663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280401 BC019397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280401 NM_133986.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108472 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07501489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07501492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07501490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07501485 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07501487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26814196 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26814194 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26814197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07542701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07501500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08016384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07542695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596903 - -
          Affymetrix HT MG-430 PM Array Plate 1424068_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420125_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420124_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1420123_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596900 - -
          Mouse Genome 430 2.0 Array 1420125_at - -
          Mouse Genome 430 2.0 Array 1420124_PM_s_at - -
          Mouse Genome 430 2.0 Array 1420124_s_at - -
          Mouse Genome 430 2.0 Array 1420123_at - -
          Mouse Genome 430 2.0 Array 1424068_PM_at - -
          Mouse Genome 430 2.0 Array 1420123_PM_at - -
          Mouse Genome 430 2.0 Array 1424068_at - -
          Mouse Genome 430 2.0 Array 1420125_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102791_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102791_at - -
          Affymetrix Mouse Clariom S Array PSR0900025752.mm.1 NM_133986.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025752.mm.1 TC0900003058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025751.mm.1 NM_133986.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025751.mm.1 TC0900003058.mm.1 MTA TC
          Mouse Expression Set 430 1424068_PM_at - -
          Mouse Expression Set 430 1424068_at - -
          Mouse Expression Set 430 1420125_at - -
          Mouse Expression Set 430 1420124_s_at - -
          Mouse Expression Set 430 1420124_PM_s_at - -
          Mouse Expression Set 430 1420125_PM_at - -
          Mouse Expression Set 430 1420123_at - -
          Mouse Expression Set 430 1420123_PM_at - -
          Murine Genome U74v2 Set 162100_PM_r_at - -
          Murine Genome U74v2 Set 162100_r_at - -
          Murine Genome U74v2 Set 104739_at - -
          Murine Genome U74v2 Set 104739_PM_at - -
          Mouse Genome 430A 2.0 Array 1420123_PM_at - -
          Mouse Genome 430A 2.0 Array 1420124_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1420123_at - -
          Mouse Genome 430A 2.0 Array 1424068_PM_at - -
          Mouse Genome 430A 2.0 Array 1420125_at - -
          Mouse Genome 430A 2.0 Array 1420125_PM_at - -
          Mouse Genome 430A 2.0 Array 1420124_s_at - -
          Mouse Genome 430A 2.0 Array 1424068_at - -
          Affymetrix HT Mouse Genome 430A Array 1424068_at - -
          Affymetrix HT Mouse Genome 430A Array 1420125_at - -
          Affymetrix HT Mouse Genome 430A Array 1420123_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424068_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420123_at - -
          Affymetrix HT Mouse Genome 430A Array 1420125_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420124_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1420124_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 609193 - -
          ABI Mouse Genome Survey Microarray 668885 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 105220039 - -
          Illumina mouse-6 v1.0 expression beadchip 2680687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254215 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742318 - -
          mouse-6 v1.1 (Illumina) scl00102791.2_289-S - -
          mouse-6 v1.1 (Illumina) scl35353.3_109-S - -
          Illumina Sentrix mouse-6.v1 bead array 105220039 - -
          Illumina Sentrix mouse-6.v1 bead array 2680687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00102791.2_289-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680687 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105220039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254215 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254215 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00102791.2_289-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline