Search
Search

Elmod2| Gene Symbol: Elmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170691.1 | NP_001164162.1 | 7 - 8 | 739 | 80 | |
| NM_178736.5 | NP_848851.3 | 7 - 8 | 744 | 80 | ||
| GenBank mRNA | AK029044.1 | - | 7 - 8 | 715 | 80 | |
| AK036746.1 | - | 7 - 8 | 684 | 80 | ||
| AK041137.1 | - | 7 - 8 | 645 | 80 | ||
| AK048572.1 | - | 3 - 4 | 1950 | 80 | ||
| AK082730.1 | - | 6 - 7 | 511 | 80 | ||
| AK084950.1 | - | 7 - 8 | 744 | 80 | ||
| BC079654.1 | - | 7 - 8 | 705 | 6855187 | 80 | |
Set Membership: |
|