Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00774934_m1
          See other FBXW19 GE Assays ›
          Gene Symbol:
          Fbxw19
          Gene Name:
          F-box and WD-40 domain protein 19
          Gene Aliases:
          E330023O18
          Chromosome Location:
          Chr.9: 109478507 - 109495863 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00774934_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxw19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177703.3 NP_808371.1 4 - 5 421 102
          GenBank mRNA AK087808.1 - 4 - 5 422 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003087.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxw19

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P219377 NM_177703.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P219377 AK087808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P219377 NM_177703.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P219377 AK087808.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30822 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160625 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P219377 NM_177703.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P219377 AK087808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P219377 NM_177703.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P219377 AK087808.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26819400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06922813 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06922832 - -
          Whole Mouse Genome G4122A A_52_P219377 NM_177703.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P219377 AK087808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P219377 NM_177703.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P219377 AK087808.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26819399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26819354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26819355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26819430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08039853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26819436 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597048 - -
          Affymetrix HT MG-430 PM Array Plate 1437316_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10597036 - -
          Mouse Genome 430 2.0 Array 1437316_at - -
          Mouse Genome 430 2.0 Array 1437316_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235612_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900026047.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026047.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026047.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026050.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026050.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026050.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026051.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026051.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026051.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026045.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026045.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026045.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026053.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026053.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026053.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026043.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026043.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026043.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026052.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026052.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026052.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026049.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026049.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026049.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026042.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026042.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026042.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026046.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026046.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026046.mm.1 TC0900003087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026048.mm.1 NM_177703.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026048.mm.1 NM_177703.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026048.mm.1 TC0900003087.mm.1 MTA TC
          Mouse Expression Set 430 1437316_PM_at - -
          Mouse Expression Set 430 1437316_at - -
          Affymetrix HT Mouse Genome 430B Array 1437316_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437316_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550620 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230055 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230055 - -
          mouse-6 v1.1 (Illumina) scl35336.10.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35336.10.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230055 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230055 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline