Search
Search

Tssk5| Gene Symbol: Tssk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183099.2 | NP_898922.2 | 2 - 3 | 563 | 83 | |
| GenBank mRNA | AK007032.1 | - | 2 - 3 | 565 | 83 | |
| BC114543.1 | - | 2 - 3 | 367 | 83 | ||
Set Membership: |
|