Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833453_g1
          See other KLK1B5 GE Assays ›
          Gene Symbol:
          Klk1b5
          Gene Name:
          kallikrein 1-related peptidase b5
          Gene Aliases:
          Klk5, mGK-5
          Chromosome Location:
          Chr.7: 44216475 - 44220703 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833453_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk1b5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008456.3 NP_032482.1 3 - 4 521 198
          GenBank mRNA BC013659.1 - 3 - 4 515 4190286 198
          BC094281.1 - 3 - 4 521 4923171 198

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000795.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klk1b5

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002933 NM_008456.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101899 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106799 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002933 NM_008456.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086126 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106799 - -
          Whole Mouse Genome G4122A A_51_P395309 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121231 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138140 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107696 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06593343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06618575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06064641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06593347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552549 - -
          Affymetrix HT MG-430 PM Array Plate 1419010_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1449313_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552618 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552546 - -
          Mouse Genome 430 2.0 Array 1419010_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455977_x_at - -
          Mouse Genome 430 2.0 Array 1419010_x_at - -
          Mouse Genome 430 2.0 Array 1449313_PM_at - -
          Mouse Genome 430 2.0 Array 1455977_PM_x_at - -
          Mouse Genome 430 2.0 Array 1449313_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16622_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16622_at - -
          Affymetrix Mouse Clariom S Array PSR0700006449.mm.1 NM_008456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006449.mm.1 TC0700000795.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006446.mm.1 NM_008456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006446.mm.1 TC0700000795.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006447.mm.1 NM_008456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006447.mm.1 TC0700000795.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006445.mm.1 NM_008456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006445.mm.1 TC0700000795.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006448.mm.1 NM_008456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006448.mm.1 TC0700000795.mm.1 MTA TC
          Mouse Expression Set 430 1455977_PM_x_at - -
          Mouse Expression Set 430 1449313_PM_at - -
          Mouse Expression Set 430 1449313_at - -
          Mouse Expression Set 430 1455977_x_at - -
          Mouse Expression Set 430 1419010_x_at - -
          Mouse Expression Set 430 1419010_PM_x_at - -
          Murine Genome U74v2 Set 100060_PM_i_at - -
          Murine Genome U74v2 Set 161637_f_at - -
          Murine Genome U74v2 Set 104495_f_at - -
          Murine Genome U74v2 Set 100060_i_at - -
          Murine Genome U74v2 Set 161637_PM_f_at - -
          Murine Genome U74v2 Set 104495_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1449313_at - -
          Mouse Genome 430A 2.0 Array 1419010_x_at - -
          Mouse Genome 430A 2.0 Array 1419010_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455977_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455977_x_at - -
          Mouse Genome 430A 2.0 Array 1449313_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419010_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1449313_at - -
          Affymetrix HT Mouse Genome 430A Array 1449313_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419010_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 416756 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590139 - -
          MouseWG-6 v2 Expression BeadChip 2350528 - -
          Illumina mouse-6 v1.0 expression beadchip 2350528 - -
          Illumina mouse-6 v1.0 expression beadchip 105420707 - -
          Illumina mouse-6 v1.0 expression beadchip 6590139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232795 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224893 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224893 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255129 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232795 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731191 - -
          mouse-6 v1.1 (Illumina) scl000139.1_0-S - -
          mouse-6 v1.1 (Illumina) scl016622.5_67-S - -
          mouse-6 v1.1 (Illumina) GI_7949063-S - -
          Illumina Sentrix mouse-6.v1 bead array 6590139 - -
          Illumina Sentrix mouse-6.v1 bead array 105420707 - -
          Illumina Sentrix mouse-6.v1 bead array 2350528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000139.1_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016622.5_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105420707 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731191 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224893 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731191 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255129 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224893 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline