Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01131751_m1
          See other MAPRE2 GE Assays ›
          Gene Symbol:
          Mapre2
          Gene Name:
          microtubule-associated protein, RP/EB family, member 2
          Gene Aliases:
          AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1
          Chromosome Location:
          Chr.18: 23752333 - 23893861 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01131751_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapre2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162941.1 NP_001156413.1 4 - 5 476 73
          NM_001162942.1 NP_001156414.1 4 - 5 419 73
          NM_153058.4 NP_694698.3 3 - 4 527 73
          GenBank mRNA AI325879.1 - 4 - 5 413 73
          AK028823.1 - 4 - 5 477 73
          AK038562.1 - 3 - 4 514 73
          AK040767.1 - 3 - 4 502 73
          AK045493.1 - 3 - 4 410 73
          AK045904.1 - 3 - 4 502 73
          AK046095.1 - 3 - 4 469 73
          AK046221.1 - 3 - 4 494 73
          AK148954.1 - 3 - 4 502 73
          AK168834.1 - 5 - 6 361 73
          BC025804.1 - 3 - 4 474 5101879 73
          BC027056.1 - 3 - 4 493 5361238 73
          BC028987.1 - 3 - 4 449 5362960 73
          BC035254.1 - 3 - 4 479 5363176 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000184.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapre2

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          mitotic nuclear division
          cell division
          protein binding
          microtubule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 NM_001162941.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 NM_001162941.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 NM_001162942.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 NM_001162942.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 NM_153058.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 NM_153058.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001339.mm.1 TC1800000184.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline