Search
Search
| Gene Symbol: Tbcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025548.3 | NP_079824.2 | 4 - 5 | 885 | 130 | |
| GenBank mRNA | AK002316.1 | - | 4 - 5 | 626 | 130 | |
| AK003655.2 | - | 4 - 5 | 631 | 130 | ||
| AK010438.1 | - | 4 - 5 | 642 | 130 | ||
| BC010684.1 | - | 4 - 5 | 618 | 2648112 | 130 | |
Set Membership: |
|