| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700016335.mm.1 |
NM_001029872.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016335.mm.1 |
NM_001029872.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016335.mm.1 |
TC0700001877.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016333.mm.1 |
NM_001029872.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016333.mm.1 |
NM_001029872.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016333.mm.1 |
TC0700001877.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016334.mm.1 |
NM_001029872.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016334.mm.1 |
NM_001029872.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016334.mm.1 |
TC0700001877.mm.1 |
MTA TC |