Search
Search

Csn1s1| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 15 - 16 | 503 | 72 | |
| NM_001286016.1 | NP_001272945.1 | 16 - 17 | 527 | 72 | ||
| NM_007784.3 | NP_031810.1 | 16 - 17 | 527 | 72 | ||
| GenBank mRNA | BC002101.1 | - | 16 - 17 | 496 | 3484624 | 72 |
| BC003859.1 | - | 16 - 17 | 511 | 3486384 | 72 | |
| BC006024.1 | - | 15 - 16 | 487 | 3592625 | 72 | |
| BC007192.1 | - | 16 - 17 | 511 | 72 | ||
| BC008278.1 | - | 16 - 17 | 511 | 2812523 | 72 | |
| BC040246.1 | - | 16 - 17 | 523 | 2647887 | 72 | |
| M36780.1 | - | 15 - 16 | 466 | 72 | ||
Set Membership: |
|