| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994492 |
AK122466.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994492 |
AK135148.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994492 |
NM_001033173.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P452019 |
AK122466.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P452019 |
AK135148.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P452019 |
NM_001033173.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P452019 |
AK122466.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P452019 |
AK135148.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P452019 |
NM_001033173.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2026079 |
AK122466.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2026079 |
NM_001033173.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703465 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
765431 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156333 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
190921 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489739 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
595032 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
AB000828.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
AK141198.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
AK213086.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
BC066058.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
BC082325.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
D17393.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
NM_019392.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
U05683.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
U18342.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
U18343.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
U18933.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
X78103.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752101 |
XM_001480339.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708286 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885183 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
868851 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
332209 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798372 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154691 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
78041 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793197 |
AK147862.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793197 |
NM_001034865.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
336850 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
852697 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
121854 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
720089 |
AK083430.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
11463 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654419 |
AK050249.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
147821 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
50785 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
700884 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
246338 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
96533 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497463 |
AK170529.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731398 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187206 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
468803 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
264874 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
335191 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
762744 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
382724 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308457 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
256456 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
262705 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319672 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
869043 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57943 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
170774 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
71728 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
34541 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
124811 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
93013 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
176741 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P161066 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2026079 |
AK122466.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2026079 |
NM_001033173.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P452019 |
AK122466.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P452019 |
AK135148.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P452019 |
NM_001033173.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P452019 |
AK122466.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P452019 |
AK135148.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P452019 |
NM_001033173.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994492 |
AK122466.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994492 |
AK135148.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994492 |
NM_001033173.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P670970 |
AK018244.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P670970 |
AK122466.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P670970 |
NM_001033173.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P452019 |
AK122466.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P452019 |
AK135148.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P452019 |
NM_001033173.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P452019 |
AK122466.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P452019 |
AK135148.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P452019 |
NM_001033173.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P138917 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117812 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474866 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474871 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474869 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474870 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474867 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06045342 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474863 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06291593 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25474865 |
- |
- |