Search
Search
| Gene Symbol: Mto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026658.2 | NP_080934.2 | 6 - 7 | 1208 | 96 | |
| GenBank mRNA | AF369902.1 | - | 6 - 7 | 1198 | 96 | |
| AK019088.1 | - | 6 - 7 | 1208 | 96 | ||
| BC063256.1 | - | 6 - 7 | 1194 | 96 | ||
Set Membership: |
|