Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179063_g1
          See other LGALS4 GE Assays ›
          Gene Symbol:
          Lgals4
          Gene Name:
          lectin, galactose binding, soluble 4
          Gene Aliases:
          gal-4
          Chromosome Location:
          Chr.7: 28833609 - 28841710 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179063_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lgals4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010706.2 NP_034836.1 7 - 8 973 148
          GenBank mRNA AF026795.1 - 6 - 7 523 148
          AF510729.1 - 7 - 8 622 148
          AK145734.1 - 7 - 8 793 148
          AY044870.1 - 7 - 8 645 148
          BC011236.1 - 7 - 8 611 4205669 148
          BC021632.1 - 7 - 8 629 4218531 148
          BC030297.1 - 7 - 8 786 4986324 148
          BC094008.1 - 7 - 8 624 4986911 148
          BC141106.1 - 7 - 8 695 9055718 148
          BC145387.1 - 7 - 8 695 9053945 148
          EF017938.1 - 7 - 8 973 148
          EF017939.1 - 7 - 8 973 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000515.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lgals4

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Process(es)

          protein binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063257 BC141106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063257 BC145387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063257 EF017939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063257 AY044870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063257 BC011236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063256 AY044870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063256 BC011236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063256 BC021632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063256 BC030297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063256 BC094008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 AY044870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC011236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC021632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC030297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC094008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 AY044870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC011236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC021632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC030297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497100 BC094008.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109160 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145450 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063257 BC141106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063257 BC145387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063257 EF017939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063257 AY044870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063257 BC011236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 AY044870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC011236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC021632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC030297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC094008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 AY044870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC011236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC021632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC030297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497100 BC094008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063256 AY044870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063256 BC011236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063256 BC021632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063256 BC030297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063256 BC094008.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023817 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01490674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145450 - -
          Whole Mouse Genome G4122A A_52_P634250 BC141106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634250 BC145387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634250 EF017939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634250 AY044870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634250 BC011236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 AY044870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC011236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC021632.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC030297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC094008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 AY044870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC011236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC021632.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC030297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497100 BC094008.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136361 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111032 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06544676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06480412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06544676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06480423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551633 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551628 - -
          Affymetrix HT MG-430 PM Array Plate 1451336_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551626 - -
          Mouse Genome 430 2.0 Array 1451336_PM_at - -
          Mouse Genome 430 2.0 Array 1451336_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16855_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16855_at - -
          Affymetrix Mouse Clariom S Array PSR0700004320.mm.1 NM_010706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004320.mm.1 NM_010706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004320.mm.1 TC0700000515.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004314.mm.1 NM_010706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004314.mm.1 NM_010706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004314.mm.1 TC0700000515.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004315.mm.1 NM_010706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004315.mm.1 NM_010706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004315.mm.1 TC0700000515.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004312.mm.1 NM_010706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004312.mm.1 NM_010706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004312.mm.1 TC0700000515.mm.1 MTA TC
          Mouse Expression Set 430 1451336_PM_at - -
          Mouse Expression Set 430 1451336_at - -
          Murine Genome U74v2 Set 160099_at - -
          Murine Genome U74v2 Set 160099_PM_at - -
          Mouse Genome 430A 2.0 Array 1451336_at - -
          Mouse Genome 430A 2.0 Array 1451336_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451336_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451336_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3390338 - -
          MouseWG-6 v2 Expression BeadChip 2260215 - -
          Illumina mouse-6 v1.0 expression beadchip 3390338 - -
          Illumina mouse-6 v1.0 expression beadchip 2260215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2626235 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2626235 - -
          mouse-6 v1.1 (Illumina) scl32857.8.1_60-S - -
          mouse-6 v1.1 (Illumina) scl016857.8_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260215 - -
          Illumina Sentrix mouse-6.v1 bead array 3390338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32857.8.1_60-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016857.8_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390338 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2968211 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2626235 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2626235 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2968211 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl016857.8_15-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline