Search
Search

Smek2| Gene Symbol: Smek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134034.2 | NP_598795.2 | 10 - 11 | 1912 | 94 | |
| GenBank mRNA | AK037253.1 | - | 10 - 11 | 1844 | 94 | |
| AK041513.1 | - | 10 - 11 | 1857 | 94 | ||
| AK044742.1 | - | 10 - 11 | 1855 | 94 | ||
| AK090014.1 | - | 10 - 11 | 1888 | 94 | ||
| AK129344.1 | - | 10 - 11 | 1823 | 94 | ||
| AK132660.1 | - | 10 - 11 | 1434 | 94 | ||
| AK166627.1 | - | 10 - 11 | 1839 | 94 | ||
| BC006870.1 | - | 10 - 11 | 1823 | 3595439 | 94 | |
Set Membership: |
|