Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01181327_m1
          See other H2AFV GE Assays ›
          Gene Symbol:
          H2afv
          Gene Name:
          H2A histone family, member V
          Gene Aliases:
          C530002L11Rik, H2a.z-2, H2av
          Chromosome Location:
          Chr.11: 6427226 - 6444443 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01181327_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2afv
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029938.1 NP_084214.1 4 - 5 430 76
          GenBank mRNA AK021267.1 - 4 - 5 353 76
          AK155105.1 - 4 - 5 433 76
          AK168110.1 - 4 - 5 402 76
          BC028539.1 - 3 - 4 225 76
          BC147293.1 - 3 - 4 389 9056353 76
          BC147294.1 - 4 - 5 415 9056344 76
          BC147741.1 - 4 - 5 415 9056690 76
          BC147751.1 - 4 - 5 415 9056700 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002170.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099520 NM_029938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099526 BC028539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099526 NM_029938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099526 AK021267.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116904 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132924 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099520 NM_029938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099526 BC028539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099526 NM_029938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099526 AK021267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472630 BC028539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472630 NM_029938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472630 AK021267.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09323747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09529737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09743175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27568545 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10384147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384148 - -
          Affymetrix HT MG-430 PM Array Plate 1428029_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1436596_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10384145 - -
          Mouse Genome 430 2.0 Array 1428029_PM_a_at - -
          Mouse Genome 430 2.0 Array 1436596_PM_at - -
          Mouse Genome 430 2.0 Array 1428029_a_at - -
          Mouse Genome 430 2.0 Array 1436596_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77605_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77605_at - -
          Affymetrix Mouse Clariom S Array PSR1100021145.mm.1 NM_029938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021145.mm.1 NM_029938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021145.mm.1 TC1100002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021144.mm.1 NM_029938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021144.mm.1 NM_029938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021144.mm.1 TC1100002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021143.mm.1 NM_029938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021143.mm.1 NM_029938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021143.mm.1 TC1100002170.mm.1 MTA TC
          Mouse Expression Set 430 1428029_PM_a_at - -
          Mouse Expression Set 430 1428029_a_at - -
          Mouse Expression Set 430 1436596_PM_at - -
          Mouse Expression Set 430 1436596_at - -
          Murine Genome U74v2 Set 96710_at - -
          Murine Genome U74v2 Set 96710_PM_at - -
          Mouse Genome 430A 2.0 Array 1428029_a_at - -
          Mouse Genome 430A 2.0 Array 1428029_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428029_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428029_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1436596_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436596_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 472385 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770594 - -
          MouseWG-6 v2 Expression BeadChip 5220129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703385 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688347 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703385 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688347 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline